Applied Biosystems™ SimpliAmp™ Strip Plate Tray Cover
Thermo Scientific™
Applied Biosystems™ SimpliAmp™ Strip Plate Tray Cover
Ease of sample setup and reduced handling steps enabled using the SimpliAmp Strip Plate Tray Cover.
Have Questions?
Catalog NumberQuantity
A5597110
Catalog number A55971
Price (MXN)
-
Quantity:
10
Pipette and centrifuge with confidence with the Applied Biosystems™ SimpliAmp™ Strip Plate Tray Cover.
Used alongside the Thermo Scientific lysis tubes and pierceable seals to securely pipette from the Lysis Tubes and alongside the Thermo Scientific Assay Tubes, securely centrifuge with the Thermo Scientific™ SureTect™ PCR Assay Workflow enabling a simple, streamlined workflow for maximum efficiency and productivity in your laboratory.
Use the SimpliAmp Strip Plate Tray Cover to securely centrifuge SureTect PCR tubes and pipette from your SureTect lysis tubes in the SureTect workflow.
Designed to work in conjunction with the Thermo Scientific lysis and PCR tubes, SimpliAmp Strip Plate Tray and pierceable seals, the SimpliAmp Strip Plate Tray Cover enables you to streamline your SureTect workflow regardless of sample throughput.
Combined with our comprehensive portfolio of assays and world-class instrumentation you can perform food safety tests confidently knowing exactly what you are getting – quality combined with simplicity.
We are committed to enabling you to experience the true confidence in your results, right-first time, every time, and the Thermo Scientific™ SureTect™ Real-Time PCR System is continually evolving to support your laboratory and testing needs.
Specifications
For Use With (Equipment)Thermo Scientific lysis and PCR tubes, SimpliAmp Strip Plate Tray and pierceable seals
Product LineSureTect
Quantity10
Sample TypeFood
Unit SizeEach