TaqPath™ COVID-19, Flu A, Flu B, RSV Select Panel, 200 rxns - FAQs
TaqPath™ COVID-19, Flu A, Flu B, RSV Select Panel, 200 rxns - FAQs
View additional product information for TaqPath™ COVID-19, Flu A, Flu B, RSV Select Panel - FAQs (A56124, A56123)
1 product FAQs found
Can I use Diomni Software v4.2 with the TaqPath COVID-19, Flu A, Flu B, RSV Select Panel IVD test (Cat. No. A56123, A56124)?
No, at this time, only Diomni Software v4.1 is approved for use with the TaqPath COVID-19, Flu A, Flu B, RSV Select Panel IVD test.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.