Can I use Diomni Software v4.2 with the TaqPath COVID-19, Flu A, Flu B, RSV Select Panel IVD test (Cat. No. A56123, A56124)?
No, at this time, only Diomni Software v4.1 is approved for use with the TaqPath COVID-19, Flu A, Flu B, RSV Select Panel IVD test.