POROS™ CaptureSelect™ FcXP Affinity Resin
Thermo Scientific™

POROS™ CaptureSelect™ FcXP Affinity Resin

Thermo Scientific™ POROS™ CaptureSelect™ FcXP Affinity Resin is specifically designed for the purification of recombinant human IgG, Fc fusion proteins, and plasma-derived IgG through binding of the CH3 domain of human IgG.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A562521000 mL
A5624950 mL
A56251250 mL
A562535 L
A5625410 L
Catalog number A56252
Price (MXN)
-
Quantity:
1000 mL

POROS CaptureSelect FcXP Affinity Resin is specifically designed for the purification of recombinant human IgG, Fc fusion proteins, and plasma-derived IgG through binding of the CH3 domain of human IgG. The FcXP ligand recognizes all human IgG subclasses (IgG1, 2, 3, and 4) with high binding capacity (20–25 g IgG/L) and excellent elution characteristics. The large pore structure of the POROS bead enables efficient purification of larger biomolecules.

Features of FcXP affinity matrix include:

  • Mild elution, making it suitable for Fc fusion proteins
  • One-step purification from crude material with high purity and yield
  • High specificity and capacity at higher flow rates
  • Excellent scalability
  • Non-animal derived

POROS CaptureSelect FcXP Affinity Resin is intended for the purification of IgG directly from plasma and Fc fusion proteins from other complex source materials. The resin enables a robust and efficient purification process with excellent purity obtained in a single step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Matrix: cross-linked poly(styrene-divinylbenzene) POROS beads
  • Volume mean particle size: ~45 μm
  • Ligand: single-domain (VHH) antibody fragment
  • Binding capacity: 20–25 g IgG/L resin independent of flow rate, column height, and contact time
  • Elution conditions: 20 mM acetic acid, pH 4
  • Mechanical resistance: 100 bar (1450 psi, 10 MPa)
  • Formulation buffer: 18% (v/v) ethanol
For Research and Development Use and Further Manufacturing in support of FDA-regulated end uses. Not for diagnostic use or direct administration to humans or animals.
Specifications
Quantity1000 mL
Shipping ConditionWet ice
FormResin Slurry
Particle Size∼ 45 μm
Product LinePOROS CaptureSelect
TypeResin
Unit SizeEach