Ion 560™ Chip Kit
Ion Torrent™

Ion 560™ Chip Kit

Ion 560™ Chip Kit for next-generation sequencing expands sequencing capacity on the Ion GeneStudio™ S5 Plus and S5 Prime systems, generating 240–280 million reads per chip for 200-base-read libraries.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A564088 Chips
A564074 Chips
Catalog number A56408
Price (USD)
20,000.00
Each
Quantity:
8 Chips
Price (USD)
20,000.00
Each

Ion 560™ Chip Kit for next-generation sequencing expands sequencing capacity on the Ion GeneStudio™ S5 Plus and S5 Prime systems, generating 240–280 million reads per chip for 200-base-read libraries. The kit includes four (Cat. No. A56407) or eight (Cat. No. A56408) barcoded semiconductor sequencing chips for sequencing with supported Ion Torrent™ assays. Use the Ion 560™ Chip Kit for supported research applications, including comprehensive genomic profiling, transcriptome profiling, and targeted sequencing with Ion AmpliSeq™ standard and HD custom assays.

The Ion 560™ Workflow is designed to help labs:

  • Expand sequencing capacity with 240–280 million reads per chip
  • Sequence supported libraries with read lengths up to 200 bases
  • Scale supported Ion Chef™ and Ion GeneStudio™ workflows while maintaining the familiar Ion Torrent™ systems
  • Maintain chip-level sample tracking with an integrated chip barcode

Notes:

  • The Ion 560™ Chip is compatible with the Ion Chef™ System, Ion GeneStudio™ S5 Plus System, and Ion GeneStudio™ S5 Prime System
  • Torrent Suite™ Software v5.22 or later is required.
  • Ion Reporter™ Software v5.22 or later is required for compatible Ion Reporter™ analysis workflows.

Use this chip with the Ion 560™ Kit – Chef (Cat. No. A56411) for automated template preparation, chip loading, and sequencing.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Next-Generation Sequencing
For Use With (Equipment)Ion GeneStudio S5 Plus System, Ion GeneStudio S5 Prime System, Ion Chef System
Number of Reads per Chip240 to 280 Million (Output varies by workflow and run conditions)
Product TypeChip Kit
Quantity8 Chips
Read LengthSupports up to 200 base-read libraries
Shipping ConditionRoom Temperature
Software CompatibilityTorrent Suite Software v5.22 or later; Ion Reporter Software v5.22 or later
Software TypeInformatics Tools
Starting MaterialDNA, RNA or cfTNA
Unit SizeEach
Contents & Storage
8 Ion 560 Chips (Store at 15°C to 30°C)