Product Image
Invitrogen™

iBlot™ 3 Electrode Replacement Kit

The iBlot 3 Electrode Replacement Kit contains four electrical contacts, four white rubber seals, and eight screws for the iBlotRead more
Have Questions?
Catalog NumberQuantity
A564271 kit
Catalog number A56427
Price (USD)
870.00
Each
Quantity:
1 kit
Price (USD)
870.00
Each

The iBlot 3 Electrode Replacement Kit contains four electrical contacts, four white rubber seals, and eight screws for the iBlot 3 Western Blot Transfer Device. Please refer to the iBlot 3 Western Blot Transfer System User Guide for instructions on how to replace the electrical contacts.

About the iBlot 3 Western Blot Transfer Device
The iBlot 3 Western Blot Transfer Device is a dry transfer device that performs western blotting transfer efficiently and reliably in as few as three minutes and without the need for liquid buffers. The iBlot 3 system is compatible with both nitrocellulose and polyvinylidene difluoride (PVDF) membranes and offers equivalent or better performance compared with traditional wet-transfer methods in a fraction of the time. The iBlot 3 Western Blot Transfer Device is an integral part of the iBlot dry-blotting system, which consists of the transfer device and consumable transfer stacks that contain the required buffers and transfer membrane (nitrocellulose or PVDF).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Membrane CompatibilityNitrocellulose, PVDF
Mode of TransferDry
Quantity1 kit
For Use With (Equipment)iBlot 3
Gel CompatibilityMini- or Midi-sized Bis-Tris Gels, Tricine Gels, Tris-Acetate Gels, and Tris-Glycine Gels
Product LineiBlot 3
TypeElectrode Replacement Kit
Unit SizeEach
Contents & Storage
• Four electrical contacts
• Four white rubber seals
• Eight screws

Store at room temperature.