Fetal Bovine Serum, Premium Plus
Fetal Bovine Serum, Premium Plus
Fetal Bovine Serum, Premium Plus
Fetal Bovine Serum, Premium Plus
Gibco™

Fetal Bovine Serum, Premium Plus

Premium Plus FBS, our most defined FBS, provides consistent cell growth for sensitive cell lines requiring the lowest viral risk.
製品番号(カタログ番号)数量フォーマット熱失活Gamma-irradiated
A566950210 x 50 mLOne ShotYesNo
A566910150 mLOne ShotYesYes
A566910210 x 50 mLOne ShotYesYes
A5669201500 mLBottleYesYes
A566940150 mLOne ShotNoNo
A566940210 x 50 mLOne ShotNoNo
A566950150 mLOne ShotYesNo
A566960150 mLOne ShotNoYes
A566960210 x 50 mLOne ShotNoYes
A5669701500 mLBottleNoNo
A5669801500 mLBottleYesNo
A5669901500 mLBottleNoYes
製品番号(カタログ番号) A5669502
価格(JPY)
-
数量:
10 x 50 mL
フォーマット:
One Shot
熱失活:
Yes
Gamma-irradiated:
No

Premium Plus FBS, our most defined FBS, provides consistent cell growth for sensitive cell lines requiring the lowest viral risk. It is ideal for use in biomanufacturing processes and diagnostic cell culture research requiring the most stringent quality profile, including an extensive biochemical and hormonal profile. Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Sera category: Premium Plus

  • Use for a broad range of cell types, especially sensitive cell lines
  • Our most characterized serum with the lowest endotoxin and hemoglobin levels
  • Endotoxin level: ≤5 EU/mL
  • Hemoglobin level: ≤20 mg/dL
  • Origin: Refer to CoA for specific origin (US, Australia, New Zealand)
  • US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US

Gibco serum delivers

  • ISO 13485-certified; processed in FDA-registered facilities
  • Triple filtered at 0.1 μm
  • Gibco bottle easier to use in hood, reduces risk of contamination and helps improve consistency
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

* FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
仕様
エンドトキシン濃度≤5 EU/mL
使用対象(アプリケーション)*FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.
Gamma-irradiatedNo
グレードPremium Plus
熱失活Yes
ヘモグロビン濃度≤20 mg/dL
処理Triple-filtered, 0.1 μm
品質保持期間5 Years
出荷条件Dry Ice
種Cattle/Bovine
試験済みUp to 70 quality tests including EMA virus testing; USP/EP mycoplasma, endotoxin, performance; biochemical/hormonal profiling
USDA食品安全性検査を含むUSDA Safety Testing Performed for Mexican and Australian origins
Age胎児
原産国特定の原産国についてはCoAを参照してください*
形状凍結
フォーマットOne Shot
製品タイプFetal Bovine Serum
数量10 x 50 mL
Serum Treatment標準(滅菌ろ過済み)
無菌性Sterile
Sterilization MethodTriple-filtered, 0.1 μm
Unit SizeEach
組成および保存条件
Store at ≤-10°C.

よくあるご質問(FAQ)

What are the benefits and disadvantages of using heat-inactivated FBS in cell culture?

  1. Heating inactivates complement. Active complement can participate in cytolytic events, contract smooth muscle, release histamine from mast cells and platelets, and activate lymphocytic and macrophage cells. Applications where heat-inactivated serum is recommended include immunological studies and culturing of embryonic stem cells (ESCs), insect cells, and smooth muscle cells.
  2. Heat inactivation helps to achieve bottle-to-bottle and lot-to-lot stability by neutralizing many factors that can vary largely from lot to lot.
  3. There aren't necessarily disadvantages to heat inactivation of FBS, but there is some evidence that suggests there may be no added benefit to it unless you are carrying out immune studies.

Note: Heat inactivation is performed in a 56 degrees C water bath for 30 min with swirling every 10 min or so for heat distribution and to lower the degree of protein aggregation/flocculant precipitation. Note: If the time or temperature is exceeded, the serum may thicken to a gel. If this occurs, the serum is no longer usable. Unnecessary heat inactivation can take up time and potentially lead to wasted reagents if a mistake is made during the protocol1.

1. Pellerin, et al., Bioengineering, published in 2021.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

How much of the total protein measured in Fetal Bovine Serum (heat inactivated or otherwise) is attributed to albumin?

Our CoAs only capture the total protein content not specifically the amount attributed to albumin.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How much Fetal Bovine Serum (FBS) do I need to add to my medium?

FBS is added to culture medium at a concentration of 2-10% to provide attachment factors, nutrients, and hormones for mammalian cells, as well as to be a buffer against disruptions like pH changes and endotoxins. FBS has significant amounts of embryonic growth promoting factors like hormones, carrier proteins, and macromolecular proteins. It also has low levels of antibodies and other growth-inhibiting components. For most basal medium, 10% FBS is used. Some applications require up to 20% supplementation, so researchers need to determine what is optimal for their specific application and cell line.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How long can I store Gibco Fetal Bovine Serum at 4 degrees C?

This product can be stored at 4 degrees C for up to 4 weeks. Once the medium has been supplemented with serum, we recommend using it within 2-4 weeks.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How should I thaw Gibco Fetal Bovine Serum?

We recommend thawing the serum overnight at 4 degrees C or in a 37 degrees C water bath, removing as soon as it is thawed. Once thawed, aliquot into single-use sizes and freeze the aliquots. Each aliquot should ideally be thawed only one additional time as repeated freeze-thaw cycles are not recommended.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.