Fetal Bovine Serum, Premium Plus
Gibco™

Fetal Bovine Serum, Premium Plus

Premium Plus FBS, our most defined FBS, provides consistent cell growth for sensitive cell lines requiring the lowest viral risk.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatHeat InactivatedGamma-irradiated
A566960210 x 50 mLOne ShotNoYes
A566910150 mLOne ShotYesYes
A566910210 x 50 mLOne ShotYesYes
A5669201500 mLBottleYesYes
A566940150 mLOne ShotNoNo
A566940210 x 50 mLOne ShotNoNo
A566950150 mLOne ShotYesNo
A566950210 x 50 mLOne ShotYesNo
A566960150 mLOne ShotNoYes
A5669701500 mLBottleNoNo
A5669801500 mLBottleYesNo
A5669901500 mLBottleNoYes
Catalog number A5669602
Price (USD)
1,114.65
Online exclusive
1,358.00
Save 243.35
Each
Quantity:
10 x 50 mL
Format:
One Shot
Heat Inactivated:
No
Gamma-irradiated:
Yes
Price (USD)
1,114.65
Online exclusive
1,358.00
Save 243.35
Each

Premium Plus FBS, our most defined FBS, provides consistent cell growth for sensitive cell lines requiring the lowest viral risk. It is ideal for use in biomanufacturing processes and diagnostic cell culture research requiring the most stringent quality profile, including an extensive biochemical and hormonal profile. Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Sera category: Premium Plus

  • Use for a broad range of cell types, especially sensitive cell lines
  • Our most characterized serum with the lowest endotoxin and hemoglobin levels
  • Endotoxin level: ≤5 EU/mL
  • Hemoglobin level: ≤20 mg/dL
  • Origin: Refer to CoA for specific origin (US, Australia, New Zealand)
  • US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US

Gibco serum delivers

  • ISO 13485-certified; processed in FDA-registered facilities
  • Triple filtered at 0.1 μm
  • Gibco bottle easier to use in hood, reduces risk of contamination and helps improve consistency
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

* FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Specifications
For Use With (Application)*FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.
Gamma-irradiatedYes
Heat InactivatedNo
ProcessingTriple filtered at 0.1 μm
Purity or Quality GradePremium Plus
USDA Safety Testing IncludedUSDA Safety Testing Performed for Mexican and Australian origins
AgeFetal
Country of OriginRefer to CoA for specific origin*
FormFrozen
FormatOne Shot
Product TypeFetal Bovine Serum
Quantity10 x 50 mL
Serum TreatmentGamma-irradiated
SterilitySterile
Sterilization MethodTriple-filtered, 0.1 μm
Unit SizeEach