UltraPure™ 5M NaCl
Invitrogen™

UltraPure™ 5M NaCl

UltraPure 5 M NaCl is a filtered, pre-mixed solution containing 292.2 g/L NaCl. This is a common stock solution usedRead more
Have Questions?
Catalog NumberQuantity
A570064 L
Catalog number A57006
Price (USD)
244.00
Each
Quantity:
4 L
Price (USD)
244.00
Each
UltraPure 5 M NaCl is a filtered, pre-mixed solution containing 292.2 g/L NaCl. This is a common stock solution used in many molecular biology and forensic applications.
For Research Use Only.
Specifications
Chemical Name or MaterialSodium Chloride
Concentration5 M
For Use With (Application)Forensics, Molecular Biology
PurityMolecular Biology Grade
Quantity4 L
Shipping ConditionRoom Temperature
Unit SizeEach