CellMask™ Deep Red Actin Tracking Stain, 1 x 60 Assays - FAQs
CellMask™ Deep Red Actin Tracking Stain, 1 x 60 Assays - FAQs
View additional product information for CellMask™ Deep Red Actin Tracking Stain - FAQs (A57248, A57245)
1 product FAQs found
What dyes are used to make the CellMask stains?
The proprietary fluorescent dyes in the CellMask stains are general cytoplasmic stains. They are not found to bind to any specific cellular component.