CellMask™ Green Actin Tracking Stain
Invitrogen™

CellMask™ Green Actin Tracking Stain

Stain and track intracellular F-actin with CellMask Green Actin Tracking Stain in live or fixed cells ICC, IHC, and 3D cell culture. The stain easily permeates live cell membranes, is retained within the cells after loading, and shows no detectable effects on viability after 24 hours of incubation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A572465 x 60 Assays
A572431 x 60 Assays
Catalog number A57246
Price (USD)
860.65
Online exclusive
896.00
Save 35.35
Each
Quantity:
5 x 60 Assays
Price (USD)
860.65
Online exclusive
896.00
Save 35.35
Each

Stain and track intracellular F-actin with CellMask Green Actin Tracking Stain in live or fixed cells in immunofluorescence (IF), immunocytochemistry (ICC), and immunohistochemistry (IHC) protocols, and in 3D cell culture. The CellMask Green Actin Tracking Stain fluorescently labels polymerized/filamentous actin (F-actin) in live or fixed cells because it is designed to readily permeate live cells, resulting in bright and uniform labeling. CellMask Green Actin Tracking Stain can be used to track F-actin in live cells for 24 hours or more. Live cells stained with CellMask Green Actin Tracking Stain can be fixed; the stain can also be used on already fixed cells. Cell Mask Green Actin Tracking Stain can be multiplexed with antibodies in IF, ICC, and IHC protocols, and can also be used in 3D cell culture for 3D imaging.

Features of Cell Mask Green Actin Tracking Stain include:

  • Stains F-actin in live or fixed cells for fluorescence 2D monolayer or 3D imaging experiments
  • Can be multiplexed with antibodies in IF/ICC/IHC experiments
  • Fluorescence maxima (Excitation/Emission): 503/512 nm; can be detected with FITC/GFP filter

CellMask actin tracking stains are available in three colors: green (Cat. No. A57243, A57246) detected with a FITC/GFP filter (Ex 503 nm/Em 512 nm), orange (Cat. No. A57244, A57247), detected with a TRITC/RFP filter (Ex 545 nm/Em 570 nm), and deep red (Cat. No. A57245, A57248) detected with a CY5/Deep Red filter (Ex 652 nm/Em 669 nm).

CellMask actin tracking stains detect F-actin with a targeting structure that closely resembles jasplakinolide. They do not label globular actin (G-actin) or monomer actin (M-actin). CellMask actin tracking stains easily pass through live cell membranes and are retained within live cells after loading. As demonstrated in the figures below, incubation with CellMask actin tracking stains has no detectable effect on viability of various cell lines after 24 hours of incubation.

CellMask actin tracking stains detect F-actin in live cells or tissue, which can then be fixed. In addition, CellMask actin tracking stains can be used to detect F-Actin in paraformaldehyde fixed cells or tissue (freshly fixed or cryo frozen only) and can be multiplexed with antibody detection. CellMask actin tracking stains can also be used in 3D cell culture for 3D imaging.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
ColorGreen
DescriptionCellMask™ Green Actin Tracking Stain
Detection MethodFluorescence Microscopy
Dye TypeFITC/GFP
Emission512 nm
Excitation Wavelength Range503 nm
For Use With (Equipment)Fluorescence Microscope, High Content Analyzer
Product LineCellMask
Quantity5 x 60 Assays
Shipping ConditionRoom Temperature
TargetF-Actin
Label TypeFluorescent Dye
Product TypeActin
Sub Cellular LocalizationActin, Cytoskeleton
Unit SizeEach
Contents & Storage
Contains 5 vials with enough reagent to stain 600 slides, when 500 μL of 1X staining solution is used per slide.

• Add 60 μL/vial of anhydrous DMSO to make a 1000X (1 mM) stock solution
• Dissolve stock solution 1:1000 in cell imaging solution to make 1X (1 μM) staining solution

Store at room temperature. Can be stored to –20°C with no freeze/thaw cycles. When stored as instructed, these reagents are effective for a minimum of 6 months after receiving.

Frequently asked questions (FAQs)

What dyes are used to make the CellMask stains?

The proprietary fluorescent dyes in the CellMask stains are general cytoplasmic stains. They are not found to bind to any specific cellular component.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.