CellMask™ Orange Actin Tracking Stain, 5 x 60 Assays - FAQs
CellMask™ Orange Actin Tracking Stain, 5 x 60 Assays - FAQs
View additional product information for CellMask™ Orange Actin Tracking Stain - FAQs (A57244, A57247)
1 product FAQs found
What dyes are used to make the CellMask stains?
The proprietary fluorescent dyes in the CellMask stains are general cytoplasmic stains. They are not found to bind to any specific cellular component.