CellMask™ Actin Tracking Stain Variety Pack - FAQs
CellMask™ Actin Tracking Stain Variety Pack - FAQs
View additional product information for CellMask™ Actin Tracking Stain Variety Pack - FAQs (A57249)
1 product FAQs found
What dyes are used to make the CellMask stains?
The proprietary fluorescent dyes in the CellMask stains are general cytoplasmic stains. They are not found to bind to any specific cellular component.