Product Image
Applied Biosystems™
MicroSEQ™ ID 16s rDNA 500 Supplemental Library v2022
The MicroSEQ ID 16S rDNA 500 Supplemental Library v2022 is a stand-alone library that extends the size of the MicroSEQ ID 16S rDNA 500 Library to strengthen the identification process when using MicroSEQ ID Microbial Identification Software.
Have Questions?
Catalog NumberQuantity
A573131 Ea.
Catalog number A57313
Price (MXN)
-
Quantity:
1 Ea.
The MicroSEQ ID 16S rDNA 500 Supplemental Library v2022 is a stand-alone library that extends the size of the MicroSEQ ID 16S rDNA 500 Library to strengthen the identification process when using MicroSEQ ID Microbial Identification Software. The Supplemental Library contains 7,138 16S rDNA public sequences of type and non-type strains of bacterial species that underwent the same rigorous validation process as the sequences in the MicroSEQ ID 16S rDNA 500 Library. The Supplemental Library is used when the MicroSEQ ID 16S rDNA 500 Library results in a percent match lower than the species ID acceptance criteria.
Key features include:
  • Quality controlled – an in-silico validated library with over 7,000 type and non-type strains quality-checked for sequence, taxonomy, nomenclature, and phylogeny
  • Simplified – reliable entries from public resources converted into a simple format for routine validation of unique environmental isolates
  • Consistent – the same acceptance criteria for analysis and the same reporting format in the software
  • Maintained – regular updates for entries and information supported by experienced Thermo Fisher Scientific technical staff
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microbial Identification
For Use With (Equipment)SeqStudio Genetic Analyzers, SeqStudio Flex Genetic Analyzers
LicenseStatic
Operating SystemMicrosoft Windows 10
Product LineMicroSeq™ ID
Product TypeMicrobial Identification Software
Quantity1 Ea.
SoftwareData Collection and Analysis Software
Sequencing TypeGenome and DNA Sequencing
Unit SizeEach
Frequently asked questions (FAQs)
What are the operating system requirements for MicroSEQ ID 16s rDNA 500 Supplemental Library v2022?
The operating system requirements for MicroSEQ ID 16s rDNA 500 Supplemental Library v2022 are Windows 10, Enterprise LTSC 2018 Microsoft Corporation, 64 bit OS (Operating System), x64 Based Processor, Version-1809.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the difference between MicroSEQ ID 16s rDNA 500 Supplemental Library v2018 and v2022?
MicroSEQ ID 16s rDNA 500 Supplemental Library v2022 contains identical content to the previous generation library, MicroSEQ ID 16s rDNA 500 Supplemental Library v2018. MicroSEQ ID 16s rDNA 500 Supplemental Library v2022 is compatible with MicroSEQ ID Microbial Identification Software v4.0 whereas MicroSEQ ID 16s rDNA 500 Supplemental Library v2018 is compatible with MicroSEQ ID Microbial Identification Software v3.1.3.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the browser compatibility requirement for MicroSEQ ID 16s rDNA 500 Supplemental Library v2022?
MicroSEQ ID 16s rDNA 500 Supplemental Library v2022 is compatible with Google Chrome v108.0.5359.73.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Which Windows OS (Operating System) is MicroSEQ ID Microbial Identification Software v4.0 compatible with?
MicroSEQ ID Microbial Identification Software v4.0 is compatible with 64-bit Windows 10 OS.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What are the computer requirements for MicroSEQ ID Microbial Identification Software v4.0?
The computer requirements for MicroSEQ ID Microbial Identification Software v4.0 are:
- Intel Core i7-8700 CPU at 3.20 Ghz, with 16 GB of RAM and a minimum of 465 GB hard disk space
- WIN 10 IoT 64-bit
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.