How is CleanCap Reagent AG different from ARCA (Anti-Reverse Cap Analog) technology?
CleanCap is a new capping technology that can provide higher mRNA yields and capping efficiencies than ARCA. CleanCap contains the cap 1 structure, which is found in higher eukaryotes such as mice or humans. ARCA contains the cap 0 structure that is found in lower eukaryotes such as yeast. Cap 1 mRNA can have superior translational activities in humans compared to Cap 0 mRNAs.