Zeba™ Spin Desalting Columns, 40K MWCO, 0.5 mL, 50 Columns, 50 columns - FAQs
Zeba™ Spin Desalting Columns, 40K MWCO, 0.5 mL, 50 Columns, 50 columns - FAQs
View additional product information for Zeba™ Spin Desalting Columns and Plates, 40K MWCO, 75 μL–10 mL - FAQs (A57761, A57760, A40002476, A40002471, A57759, A57758, A57768, A57756, A57767, A57766, A57765, A57764, A57763, A57762)
2 product FAQs found
What reagents and conditions are Zeba Desalting columns, plates, and cartridges compatible with?
Zeba Desalting columns, plates, and cartridges are compatible with a range of reagents and conditions. They have been confirmed to be compatible with the following:
- pH range of 3-10
- Mild oxidants/reductants
- Chaotropes (stable in 8 M urea and 6 M guanidine HCl)
- Salts
- Alcohols up to 20%
- Organic solvents such as dimethyl sulfoxide (DMSO) and dimethylformamide (DMF) (we recommend using a step-gradient of increasing concentration of the organic solvent during column equilibration)
Find additional tips, troubleshooting help, and resources within our Protein Dialysis, Desalting, and Concentration Support Center.