Sodium Chloride (NaCl) Solutions, 0.1–2.0 M
Gibco™

Sodium Chloride (NaCl) Solutions, 0.1–2.0 M

Gibco Sodium Chloride (NaCl) solutions are sterile-filtered, packaged in bioprocess containers, and ready-to-use. Sourcing NaCl solutions can help streamline your buffer preparation allowing you to focus on additional critical steps in your biopharmaceutical processes.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량포장농도
A580280220 LTote0.2 M
A57922015 LSample0.1 M
A579220220 LTote0.1 M
A5792203200 LDrum0.1 M
A58028015 LSample0.2 M
A5802803200 LDrum0.2 M
A58029015 LSample0.5 M
A580290220 LTote0.5 M
A5802903200 LDrum0.5 M
A58030015 LSample1.0 M
A580300220 LTote1.0 M
A5803003200 LDrum1.0 M
A58031015 LSample2.0 M
A580310220 LTote2.0 M
A5803103200 LDrum2.0 M
카탈로그 번호 A5802802
제품 가격(KRW)
829,000
Each
수량:
20 L
포장:
Tote
농도:
0.2 M
제품 가격(KRW)
829,000
Each
We mix compendial-grade sodium chloride (NaCl) with the same bulk water used in our Gibco Water for Injection (WFI) for Cell Culture to produce solutions that meet the most stringent expectations of the pharmaceutical, biopharma, and diagnostic industries. NaCl solutions are manufactured to cGMP (21CFR820) and ISO 13485 standards, promoting consistency and convenience for every stage of the bioprocessing pipeline.

Product features:
• Multiple concentrations available (0.1 M, 0.2 M, 0.5 M, 1.0 M, 2.0 M)
• Available in a range of pack sizes (5 L, 20 L, 200 L)
• Established products with set specifications are ready to order
• Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
• Readily accessible documentation for accelerated qualification

User benefits:
• Eases the time requirements dedicated for in-house buffer preparation
• Mitigates space required for equipment storage
• Streamlines in-house chromatography processes related to formulation, hydration, testing, and validation
• Meets workflow needs for all stages of biologic development and manufacturing

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
사양
CAS7647-14-5
인증/적합성GMP - 21 CFR 820, ISO 13485
화학물질 이름 또는 재질Sodium Chloride
색상Clear
엔도톡신 수준≤ 1.0 EU/mL
제제0.2 M NaCl
삼투압354–394 mOsm/kg
포장Tote
유통 기한12 Months
멸균Sterile-filtered
멸균 방법0.2 μm Sterile-filtered
농도0.2 M
용도(애플리케이션)Bioproduction
등급Compendial
분자량(g/mol)58.44
물리적 형태Liquid
수량20 L
용액 유형Buffer
pH4.5–8.0
Unit SizeEach