Sodium Chloride (NaCl) Solutions, 0.1–2.0 M
Gibco™

Sodium Chloride (NaCl) Solutions, 0.1–2.0 M

Gibco Sodium Chloride (NaCl) solutions are sterile-filtered, packaged in bioprocess containers, and ready-to-use. Sourcing NaCl solutions can help streamline your buffer preparation allowing you to focus on additional critical steps in your biopharmaceutical processes.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidadEnvaseConcentración
A5803003200 LBidón1,0 m
A57922015 LMuestra0,1 M
A579220220 LCaja0,1 M
A5792203200 LBidón0,1 M
A58028015 LMuestra0,2 M
A580280220 LCaja0,2 M
A5802803200 LBidón0,2 M
A58029015 LMuestra0,5 M
A580290220 LCaja0,5 M
A5802903200 LBidón0,5 M
A58030015 LMuestra1,0 m
A580300220 LCaja1,0 m
A58031015 LMuestra2,0 M
A580310220 LCaja2,0 M
A5803103200 LBidón2,0 M
Número de catálogo A5803003
Precio (MXN)
-
Cantidad:
200 L
Envase:
Bidón
Concentración:
1,0 m
We mix compendial-grade sodium chloride (NaCl) with the same bulk water used in our Gibco Water for Injection (WFI) for Cell Culture to produce solutions that meet the most stringent expectations of the pharmaceutical, biopharma, and diagnostic industries. NaCl solutions are manufactured to cGMP (21CFR820) and ISO 13485 standards, promoting consistency and convenience for every stage of the bioprocessing pipeline.

Product features:
• Multiple concentrations available (0.1 M, 0.2 M, 0.5 M, 1.0 M, 2.0 M)
• Available in a range of pack sizes (5 L, 20 L, 200 L)
• Established products with set specifications are ready to order
• Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
• Readily accessible documentation for accelerated qualification

User benefits:
• Eases the time requirements dedicated for in-house buffer preparation
• Mitigates space required for equipment storage
• Streamlines in-house chromatography processes related to formulation, hydration, testing, and validation
• Meets workflow needs for all stages of biologic development and manufacturing

Para su uso en investigación o procesos de fabricación posteriores. No apto para uso diagnóstico ni para la administración directa en seres humanos ni en animales.
Especificaciones
CAS7647-14-5
Certificaciones/conformidadGMP - 21 CFR 820, ISO 13485
Nombre del producto químico o materialSodium Chloride
ColorClear
Nivel de endotoxinas≤ 1.0 EU/mL
Formulación1.0 M NaCl
Osmolalidad1810–1937 mOsm/kg
EnvaseBidón
Duración de almacenamiento12 Months
EsterilidadSterile-filtered
Método de esterilización0.2 μm Sterile-filtered
Concentración1,0 m
Para utilizar con (aplicación)Bioproduction
GradoCompendial
Peso molecular (g/mol)58.44
Forma físicaLiquid
Cantidad200 L
Tipo de soluciónBuffer
pH4.5–8.0
Unit SizeEach