Ethanol (EtOH), 20%
Gibco™

Ethanol (EtOH), 20%

Gibco 20% Ethanol (EtOH) is sterile-filtered, packaged in bioprocess containers, and stocked to be available when you need it. Sourcing 20% EtOH can streamline your workload for equipment sanitization and column storage, allowing you to focus on the key steps in your downstream processes.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
20 L
200 L
Catalog number A5803202
Price (USD)
638.00
Each
Quantity:
20 L
Price (USD)
638.00
Each

Ethanol (EtOH) and bulk Water for Injection (WFI) are combined to produce a high-quality 20% ethanol solution designed for downstream bioprocessing applications, including chromatography column and resin storage as well as equipment sanitization. Gibco™ 20% Ethanol is manufactured using multicompendial-grade raw materials in an ISO-certified, cGMP-compliant facility to meet the stringent requirements of pharmaceutical and biopharmaceutical manufacturing.

Each lot is 0.2 μm sterile filtered and supported by a lot-specific Certificate of Analysis (CoA) to ensure consistency, traceability, and confidence in regulated workflows. With defined specifications and ready-to-use formats, Gibco™ 20% Ethanol helps reduce variability and supports reliable process performance across development and manufacturing.

  • Supports reliable chromatography resin and column storage by maintaining microbial control and preserving system readiness between runs
  • Reduces risk of in-house preparation errors by providing a ready-to-use, quality-controlled ethanol solution
  • Enables faster turnaround between batches by simplifying cleaning and storage workflows
  • Supports audit readiness and regulatory compliance with defined specifications and lot-specific documentation (CoA)
  • Minimizes handling of high-proof ethanol on site, improving safety and operational efficiency
  • Improves process consistency by eliminating variability associated with manual dilution and preparation
  • 24 months shelf life from date of manufacture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Certifications/ComplianceGMP - 21 CFR 820, ISO 13485
Chemical Name or MaterialEthyl Alcohol
ColorClear
DOT InformationDOT Transport Hazard Class 3, Packaging Group III
Formulation20% EtOH
PackagingTote
Physical FormLiquid
Shelf Life24 Months
Sterilization Method0.2 μm Sterile-filtered
Quantity20 L
Unit SizeEach