Ethanol (EtOH), 20%
Gibco™

Ethanol (EtOH), 20%

Gibco 20% Ethanol (EtOH) is sterile-filtered, packaged in bioprocess containers, and stocked to be available when you need it. Sourcing 20% EtOH can streamline your workload for equipment sanitization and column storage, allowing you to focus on the key steps in your downstream processes.
Have Questions?
Change viewbuttonViewtableView
Packaging:
Drum
Sample
Tote
Quantity:
View all
200 L
Catalog number A5803203
Price (USD)
2,975.00
Each
Packaging:
Drum
Quantity:
200 L
Price (USD)
2,975.00
Each
Ethanol (EtOH) and bulk Water for Injection (WFI) are mixed to produce solutions that meet the most stringent quality control expectations in accordance with the pharmaceutical, biopharma, and diagnostic industries. Gibco 20% Ethanol is manufactured to cGMP standards at an ISO certified facility, promoting quality and convenience for every stage of a biologic’s pipeline.

Product features:
• Available in a range of pack sizes (5 L, 20 L, 200 L)
• Product specifications are well-defined, with the established solutions ready to order
• Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
• Readily accessible documentation for accelerated qualification

User benefits:
• Meets workflow needs for all stages of biologic development and manufacturing
• Streamlines in-house processes related to formulation, hydration, testing, and validation
• Mitigates space required for buffer preparation and storage
• Eases the time requirements dedicated for in-house buffer preparation

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Certifications/ComplianceGMP - 21 CFR 820, ISO 13485
Chemical Name or MaterialEthyl Alcohol
ColorClear
Concentration20%
DOT InformationDOT Transport Hazard Class 3, Packaging Group III
Formulation20% EtOH
PackagingDrum
Physical FormLiquid
Shelf Life12 Months
Sterilization Method0.2 μm Sterile-filtered
Quantity200 L
Unit SizeEach