TrueMark™ Enteric Viral Select Panel Combo Kit
TrueMark™ Enteric Viral Select Panel Combo Kit
Applied Biosystems™

TrueMark™ Enteric Viral Select Panel Combo Kit

The TrueMark Enteric Viral Select Panel Combo Kit includes the TrueMark Enteric Viral Select Panel and TrueMark Infectious Disease 1‐StepRead more
Have Questions?
Catalog NumberQuantity
A58079200 reactions
Catalog number A58079
Price (USD)
3,875.00
Each
Add to cart
Quantity:
200 reactions
Price (USD)
3,875.00
Each
Add to cart

The TrueMark Enteric Viral Select Panel Combo Kit includes the TrueMark Enteric Viral Select Panel and TrueMark Infectious Disease 1‐Step Multiplex Master Mix for simplified ordering. Together with the TrueMark Enteric Viral Select Amplification Control (offered separately), the kit can be used to quickly evaluate and research up to 200 samples consisting of extracted nucleic acid from stool specimens.

The kit is recommended for use with the QuantStudio 5 Real-Time PCR System (with 96-well, 0.2-mL block). It includes primers and probe sets specific to the following targets:
• Rotavirus A
• Astrovirus
• Adenovirus F40/41
Bacillus atrophaeus (used as a process control)

Features of the TrueMark Enteric Viral Select Panel Combo Kit include:
• Detects and differentiates targets found in enteric viral infections
• Affordable and scalable—add to existing infrastructure to expand research menu while maintaining low operational costs
• Ready-to-use off-the-shelf solution—seamlessly integrate test into existing workflow

The TrueMark Enteric Viral Select Panel Combo Kit includes the following components:
TrueMark Enteric Viral Select Panel—multiplexed assay that contains primer and probe sets
TrueMark Infectious Disease 1‐Step Multiplex Master Mix (No ROX)—ready-to-use PCR mix, including polymerase, dNTPs, salt, and buffer

Use this kit with:
TrueMark Enteric Parasitic Select Amplification Control—plasmid control containing target-specific templates

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Research Use Only.
Specifications
No. of Reactions200 reactions
Quantity200 reactions
Detection MethodqPCR
For Use With (Application)Pathogen Detection, Real Time PCR (qPCR), RT-PCR
FormLiquid
PCR MethodMultiplex qPCR, qPCR, RT-PCR
Reaction Speed120 min.
Unit SizeEach