CTS™ Compleo™ Single-Use Kit
Gibco™

CTS™ Compleo™ Single-Use Kit

The Gibco CTS Compleo Single-Use Kit is part of the CTS Compleo Fill and Finish System. The Single-Use Kit supports automated, closed-system processing for formulation and fill workflow steps.
Have Questions?
Catalog NumberQuantity
A581655 Kits
Catalog number A58165
Price (USD)
-
Quantity:
5 Kits

The Gibco CTS Compleo Single-Use Kit is part of the CTS Compleo Fill and Finish System. The Single-Use Kit supports automated, closed-system processing for formulation and fill workflow steps.

Features of the CTS Compleo Single-Use Kit include:

  • Gamma-irradiated, individually packaged, sterile kit
  • 11 user-configurable lines that support attachment through sterile welding or luer lock connection
  • 1 L mixing bag supports volumes up to 750 mL for formulation and filling
  • Kit design reduces residual volume by minimizing the wetted surface area and purging the lines with sterile air after fluid transfer

The CTS Compleo Single-Use Kit provides flexibility for process development and readily supports commercial-scale manufacturing. Cryobags, vials, filters, and other accessories can be easily attached to the Single-Use Kit via sterile welding or luer lock connection.

Tubing material: DEHP-free PVC

Tube dimensions: 4 mm OD, 2.4 mm ID

Pump tubing: PharmaPure

Note: Kits are gamma-irradiated and individually packaged.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Certifications/ComplianceISO 13485
DimensionsTubing: 3/32 in. I.D., 5/32 in. O.D.
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
For Use With (Equipment)CTS Compleo Fill and Finish System
Product TypeSingle-Use Consumable
Quantity5 Kits
SterilitySterile
Sterilization MethodGamma-irradiated
Tubing Line 111 weldable tubes - up to 4 inputs and 10 outputs
Tubing Material Line 1PVC
Working Volume750 mL
Capacity (Metric)1 L
Tubing StylePVC
Unit SizeEach