Diomni™ Software for Diomni Enterprise IVD
Applied Biosystems™
Diomni™ Software for Diomni Enterprise IVD
Diomni Software streamlines workflows by digitally integrating real-time PCR (qPCR) systems for routine testing and test development and reducing data review time with results interpretation and automated QC checks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLicense
A581951 Year
A583412 Years
A400073663 Years
A400073655 Years
Catalog number A58195
Price (USD)
-
License:
1 Year
Applied Biosystems Diomni Software is an advanced, browser-based application designed to digitally integrate real-time PCR (qPCR) systems. It supports the planning of an extraction run, running of a qPCR test, reviewing of interpretive results and QC checks, and releasing of reports. Diomni Software can be deployed on a local network to enable centralized data management and role-based access.
Purchase includes an Enterprise license that unlocks additional features. This license must be renewed at the end of the licensing period to maintain access to the Enterprise features.
The software is US-IVD and IVDR compliant.
Features of Diomni Software include:
  • Advanced results interpretation capabilities
    • Comprehensive automated quality control (QC) checks
    • Integrated sample and reagent tracking library
    • Connectivity to laboratory information systems (LIS) and laboratory information management systems (LIMS) for efficient test order reception and result transmission
  • Connect and start runs on the QuantStudio 7 Pro Dx and QuantStudio 5 Dx real-time PCR systems and receive results automatically after run completion
  • Interface with KingFisher Apex Dx Sample Purification System to send extraction runs and receive status updates in real time
  • Scalable user management supporting an unlimited number of users
  • Enhanced data management and workflow optimization tools, including automated QC checks, sample tracking, and LIS/LIMS integration
Advanced run templates for standardized testing
Diomni Software offers advanced run templates (ADF) that significantly enhance standardization and reduce user errors across multiple sites and laboratories. These templates allow for the locking of test parameters, ensuring consistency among various users.
The software supports a wide range of applications, including presence/absence, standard curve, relative quantification, melt, and genotyping. Furthermore, the system allows for the configuration of results interpretation rules, enabling precise sample result determination. Advanced options such as delta Cq for grouped targets in presence/absence tests, definition of Cq ranges for grey zones, and quality control settings based on curve quality provide users with comprehensive analytical capabilities.
Centralized installation for efficient fleet management
The Diomni Software installation process is designed for optimal efficiency and accessibility. A one-time on-site setup in a central location on a local area network (LAN) enables seamless fleet management. This configuration allows users to initiate qPCR runs on multiple networked instruments simultaneously. Notably, there is no limitation on the number of networked users who can access the shared database across the lab network, facilitating collaborative research and streamlined operations.
Comprehensive workflow support
Diomni Software provides extensive workflow support, guiding users through every stage of the experimental process. The software assists in planning extraction runs, executing qPCR tests, reviewing interpretive results and quality control checks, and generating and releasing reports. This end-to-end support ensures a smooth and efficient experimental workflow, reducing the potential for errors and increasing overall productivity.
Enhanced functionality with internet connectivity
While Diomni Software operates effectively without an internet connection, additional features become available when connected. These include access to a reagent database for document downloads, automated license renewal, and the ability to contact remote technical support through Smart Help. For a comprehensive overview of configuration options, users are encouraged to refer to the connectivity brochure. This flexibility allows laboratories to tailor their setup according to their specific needs and security requirements.
For In Vitro Diagnostic Use
Specifications
License1 Year
Quantity1 Ea.
Software CompatibilityQuantStudio 7 Pro Dx and QuantStudio 5 Dx, Windows 11 or Windows 10, SAE Administrator Console v3 or SAE Administrator Console Dx v1.2
Product TypeSoftware
Unit SizeEach
Frequently asked questions (FAQs)
Can I see the amplification plots in Diomni Software once a run starts?
No. The amplification plots will not display in real-time. During the run, you will see the instrument status and the run time remaining on the experiment.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Can I prevent a user from omitting wells from a completed run in Diomni Software?
No, this is not supported with the current release of Diomni Software.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
When using Diomni Software with the Presence/Absence application, why are the results colored with red used for positive and green for negative?
Red is used to display positive test results in the Presence/Absence application because molecular diagnostic tests routinely screen for pathogens. The red is used as a warning flag to draw attention to a positive test result.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Where and when can I use a barcode scanner to input reagent information with Diomni Software?
An external barcode scanner acts as a keyboard emulator and can be used to enter sample IDs, plate IDs, and reagents. Thermo Fisher Scientific reagent barcodes are designed to allow population of reagent name and part number when setting up an extraction or qPCR run. The user may enter lot number and expiration date manually.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Can I use expired reagents with Diomni Enterprise Software IVD? Will expired reagents be flagged?
In Diomni Enterprise IVD Software, expired reagents will display a warning in the RUO workspace for extraction and qPCR runs and will be noted on the report if reagent information is included. Expired reagents cannot be used in the IVD workspace and the software will not allow the user to save or start a run with expired reagents.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.