Diomni™ Software for Diomni Enterprise RUO, 90-day trial license - FAQs
Diomni™ Software for Diomni Enterprise RUO, 90-day trial license - FAQs
View additional product information for Diomni™ Software for Diomni Enterprise RUO - FAQs (A40007364, A58196, A58197, A40007361, A58342)
1 product FAQs found
Is there a trial option for the Diomni Software (Enterprise)?
Yes, new installations of Diomni Software will install as Diomni Software (Enterprise) for 90 days. Software upgrades do not qualify.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.