Pharma Analytics KingFisher™ Apex 96 Deep-Well Tip Combs, Barcoded
Thermo Scientific™
Pharma Analytics KingFisher™ Apex 96 Deep-Well Tip Combs, Barcoded
These tip combs are for use with the Pharma KingFisher Flex 96 Deep-Well Head Magnetic Particle Processor or MagMAX Express-96 Deep-Well Magnetic Particle Processor in combination with PrepSEQ ResDNA kits and ProteinSEQ kits.
Have Questions?
Catalog NumberQuantity
A58353BPCase of 50
Catalog number A58353BP
Price (KRW)
1,218,000
Online offer
Ends: 31-Dec-2026
1,432,000
Save 214,000
Case of 50
Quantity:
Case of 50
Price (KRW)
1,218,000
Online offer
Ends: 31-Dec-2026
1,432,000
Save 214,000
Case of 50
These tip combs are for use with the Pharma KingFisher Flex 96 Deep-Well Head Magnetic Particle Processor or MagMAX Express-96 Deep-Well Magnetic Particle Processor in combination with PrepSEQ ResDNA kits and ProteinSEQ kits.
Optimize performance using KingFisher Flex System consumables designed specifically for use with these instruments. KingFisher Flex disposable plastics (tip combs and microplates) are made of polypropylene and are ideal for magnetic particle processing because of their low binding affinity for biomolecules. Their special design enables excellent recovery of magnetic beads.
Specifications
DisposableSingle-use
QuantityCase of 50
Format96 Deep-well Comb
Product TypeTip Comb
Sample TypeTissue Homogenate
Unit SizeCase of 50