Optical-Quality Imaging Plates, 384-well, black-walled, clear bottom
Optical-Quality Imaging Plates, 384-well, black-walled, clear bottom
Optical-Quality Imaging Plates, 384-well, black-walled, clear bottom
Thermo Scientific™
Optical-Quality Imaging Plates, 384-well, black-walled, clear bottom
These imaging plates are optically transparent, extra-low, ultra-flat, 384-well plates that provide optimal performance for microscopy and plate-reader-based assays.
Have Questions?
카탈로그 번호수량
A5894132 plates
카탈로그 번호 A58941
제품 가격(KRW)
1,254,000
온라인 행사
Ends: 31-Dec-2026
1,475,000
할인액 221,000
Each
수량:
32 plates
제품 가격(KRW)
1,254,000
온라인 행사
Ends: 31-Dec-2026
1,475,000
할인액 221,000
Each
These imaging plates are optically transparent, extra-low, ultra-flat, 384-well plates that provide optimal performance for microscopy and plate-reader-based assays. The black-walled clear-bottom 384-well imaging plates come tissue-culture-treated, pre-sterilized, and labeled with unique barcodes. They are ideal for fluorescence high-content screening and imaging assays on our CellInsight High-Content Screening instruments, stand-alone microscopes such as the EVOS M7000/5000, and multimodal plate readers such as the Varioskan LUX microplate reader.
Features of these Imaging Plates include:
  • Quality results – no more out-of-focus cells. Optical-quality cyclo-olefin polymer (COP) provides best-in-class flatness (100 μm from edge to edge), transmittance, and glass-matching refractive index (1.34).
  • Extra-low base – skirt height of 0.21 mm allows for all wells and fields of view to ne read with immersion and high NA lenses
  • Faster read times – reduce autofocus search times due to the ultra-flat plate bottom. Improved transmittance of COP allows for shorter exposure times, which can reduce imaging times and minimize light exposure to the sample
  • Results you can trust – COP is ideal for fluorescence imaging assays since it is optically transparent in a broad wavelength range, along with being 100% biocompatible, chemically inert, and mechanically stable
  • Add and go – plates come tissue-culture treated and pre-sterilized, ready for immediate cell plating. Custom coatings available upon request
  • Screening ready – plates come with unique barcodes, ready for your high content screening assays. Custom barcodes available upon request
When running high-content screening and imaging assays, high image quality is essential for good results. Several factors can impair image quality, such as using thick, uneven plates, which can lead to focusing differences between wells and image quality issues across a plate. The optical properties of these imaging plates produce superior image quality and resolution from each well when used with fluorescence or luminescence readers. The flatness of the plate bottom also yields excellent performance in laser-based autofocus systems. Using these Optical-Quality Imaging Plates helps get the most from automated imaging systems and ensure that critical assays and samples are not compromised due to inferior plates.
For Research Use Only.
사양
오토클레이브 가능Not Autoclavable
일회용Single-use
뚜껑With TSP Lid
케이스당 수32 plates
수량32 plates
어레이16 x 24
바코드Alphanumeric
색상Black
Fit for PurposeSterile and PCR Ready
물질Cyclic Olefin Polymer
웰 수384
멸균Sterile
표면 처리Tissue Culture
웰 형상Flat Bottom (F-well)
Unit SizeEach