Hamilton™ CO-RE™ II Conductive Filter Tips, non-sterile, 50 μL
Thermo Scientific™

Hamilton™ CO-RE™ II Conductive Filter Tips, non-sterile, 50 μL

These non-sterile conductive filter-tips are for use on the HID NIMBUS Presto QNA and HID Nimbus QNA systems.
Have Questions?
Catalog NumberQuantity
A589781 x 5760 tips
Catalog number A58978
Price (USD)
532.00
Each
Quantity:
1 x 5760 tips
Price (USD)
532.00
Each

These non-sterile conductive filter-tips are for use on the HID NIMBUS Presto QNA and HID Nimbus QNA systems.

For Research Use Only.

Specifications
Certifications/ComplianceCertified for Biological Purity as Dnase-/Rnase-free and non-pyrogenic
DisposableSingle-use
GraduatedNo
Length (Metric)50.4 mm
Quantity1 x 5760 tips
AutoclavableNot Autoclavable
ColorBlack
Color CodeDark Gray
FilteredFiltered
Fit for PurposeForensic Ready
For Use With (Equipment)HID Nimbus Presto QNA System, HID Nimbus QNA System
FormatRack, Blister
MaterialPolypropylene
Product LineHamilton
SterilityNon-sterile
Tip StyleConductive
Volume (Metric)50 μL
Unit SizeEach