Acetic Acid Solutions
Gibco™

Acetic Acid Solutions

Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 L
20 L
200 L
Konzentration:
2 M
3 M
4 M
Katalognummer A59169BA
Preis (EUR)
-
Menge:
5 L
Konzentration:
4 M

Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.

Features

  • Concentrated for dilution on-site
  • Available in a range of pack sizes
  • Established shelf life: set specifications
  • Made to order
  • Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
  • Readily accessible documentation to help accelerate qualification

Benefits

  • Helps ease time requirements for in-house buffer preparation
  • Mitigates space constraints for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Addresses workflow needs for biologics development and manufacturing stages

Additional information

  • Multi-compendial testing: compendial-grade or equivalent raw materials
  • Primary packaging: TAegis 5-14 single-use bioprocess container (BPC)
  • Secondary packaging: rigid shipping container with appropriate dunnage
  • Manufacturing facilities: ISO 13485–certified, cGMP-compliant to 21 CFR Part 820 standard
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS64-19-7
Chemischer Name oder MaterialRaw Material: Acetic Acid Glacial
Konzentration4 M
VerpackungBioprocess Container in Secondary Packaging
Physikalische FormSolution
GüteEP/JP/USP
Molekulargewicht (g/mol)Raw Material: 60.05
Reinheit (%)Not Available
Menge5 L
Unit SizeEach