Glucose, high-temperature short-time (HTST)-treated
Gibco™

Glucose, high-temperature short-time (HTST)-treated

Gibco™ high-temperature short-time (HTST)-treated products are briefly exposed to temperatures of ≥100°C using in-line pasteurization technology.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidadConcentraciónFormato
A59343BA5 L en BPC50 % (p/v)Muestra
A59773BA5 L en BPC25 % (p/v)Muestra
A59773BB20 L en BPC25 % (p/v)Caja
A59773BC50 L en BPC25 % (p/v)Bidón
A59773BD100 L en BPC25 % (p/v)Bidón
A59773BE200 L en BPC25 % (p/v)Bidón
A59774BA5 L en BPC40 % (p/v)Muestra
A59774BB20 L en BPC40 % (p/v)Caja
A59774BC50 L en BPC40 % (p/v)Bidón
A59774BD100 L en BPC40 % (p/v)Bidón
A59774BE200 L en BPC40 % (p/v)Bidón
A59343BB20 L en BPC50 % (p/v)Caja
A59343BC50 L en BPC50 % (p/v)Bidón
A59343BD100 L en BPC50 % (p/v)Bidón
A59343BE200 L en BPC50 % (p/v)Bidón
Número de catálogo A59343BA
Precio (MXN)
-
Cantidad:
5 L en BPC
Concentración:
50 % (p/v)
Formato:
Muestra
Gibco™ high-temperature short-time (HTST)-treated products are briefly exposed to temperatures of ≥100°C using in-line pasteurization technology. HTST is risk mitigation step that may reduce potential viral contaminations from entering upstream manufacturing from raw materials. Products such as glucose and other simple sugars are heated for a controlled period to not alter the treated product’s performance or viability. If HTST is a critical step in your process, outsourcing glucose can help you discover new efficiencies to reclaim time, space, and resources.

Product Features

  • Multiple formulations and concentrations available
  • Available in a range of pack sizes (5 L, 20 L, 50 L, 100 L, and 200 L)
  • Established products with set specifications are made to order
  • Product standardization enables shorter times from inquiry to quote and shipment, compared to custom
  • Readily accessible documentation for accelerated qualification

Benefits

  • Reduces time spent preparing buffers in-house
  • Mitigates space requirement for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Meets workflow needs across stages of development and manufacturing of biologicals

Para su uso en investigación o procesos de fabricación posteriores. No apto para uso diagnóstico ni para la administración directa en seres humanos ni en animales.

Especificaciones
ColorClear, colorless solution
Concentración50 % (p/v)
Nivel de endotoxinas≤1.00
FormatoMuestra
FormulaciónGlucose, HTST-treated, 50% (w/v)
Calidad de fabricacióncGMP
Cantidad5 L en BPC
Duración de almacenamiento24 Months
Probado paraGlucose concentration
FormularioLíquido
Tipo de productoGlucosa
EsterilidadEstéril con filtro
Unit SizeEach