Glucose, high-temperature short-time (HTST)-treated
Gibco™

Glucose, high-temperature short-time (HTST)-treated

Gibco™ high-temperature short-time (HTST)-treated products are briefly exposed to temperatures of ≥100°C using in-line pasteurization technology.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityConcentrationFormat
A59343BC50L BPC50% (w/v)Drum
A59773BA5L BPC25% (w/v)Sample
A59773BB20L BPC25% (w/v)Tote
A59773BC50L BPC25% (w/v)Drum
A59773BD100L BPC25% (w/v)Drum
A59773BE200L BPC25% (w/v)Drum
A59774BA5L BPC40% (w/v)Sample
A59774BB20L BPC40% (w/v)Tote
A59774BC50L BPC40% (w/v)Drum
A59774BD100L BPC40% (w/v)Drum
A59774BE200L BPC40% (w/v)Drum
A59343BA5L BPC50% (w/v)Sample
A59343BB20L BPC50% (w/v)Tote
A59343BD100L BPC50% (w/v)Drum
A59343BE200L BPC50% (w/v)Drum
Catalog number A59343BC
Price (TWD)
-
Quantity:
50L BPC
Concentration:
50% (w/v)
Format:
Drum
Gibco™ high-temperature short-time (HTST)-treated products are briefly exposed to temperatures of ≥100°C using in-line pasteurization technology. HTST is risk mitigation step that may reduce potential viral contaminations from entering upstream manufacturing from raw materials. Products such as glucose and other simple sugars are heated for a controlled period to not alter the treated product’s performance or viability. If HTST is a critical step in your process, outsourcing glucose can help you discover new efficiencies to reclaim time, space, and resources.

Product Features

  • Multiple formulations and concentrations available
  • Available in a range of pack sizes (5 L, 20 L, 50 L, 100 L, and 200 L)
  • Established products with set specifications are made to order
  • Product standardization enables shorter times from inquiry to quote and shipment, compared to custom
  • Readily accessible documentation for accelerated qualification

Benefits

  • Reduces time spent preparing buffers in-house
  • Mitigates space requirement for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Meets workflow needs across stages of development and manufacturing of biologicals
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
ColorClear, colorless solution
Concentration50% (w/v)
Endotoxin Level≤1.00
FormatDrum
FormulationGlucose, HTST-treated, 50% (w/v)
Manufacturing QualitycGMP
Quantity50L BPC
Shelf Life24 Months
Tested ForGlucose concentration
FormLiquid
Product TypeGlucose
SterilitySterile-filtered
Unit SizeEach