HBSS, no calcium, no magnesium, no phenol red
Gibco™

HBSS, no calcium, no magnesium, no phenol red

Gibco™ Balanced Salt Solutions are sterile-filtered, packaged in bioprocess containers, and ready to order.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
A59772BA5 LSample
A59772BB20 LTote
A59772BC50 LDrum
A59772BD100 LDrum
A59772BE200 LDrum
Catalog number A59772BA
Price (USD)
-
Quantity:
5 L
Format:
Sample
Gibco™ Balanced Salt Solutions are sterile-filtered, packaged in bioprocess containers, and ready to order. Commonly used for processing cells and flushing equipment, sourcing Gibco balanced salt solutions can help streamline your operation by reducing buffer preparation and allowing you to focus on additional critical steps in your biopharmaceutical processes. Discover new outsourcing efficiencies to reclaim time, space, and resources.

Product Features

  • Multiple formulations and concentrations available
  • Available in a range of pack sizes (5 L, 20 L, 50 L, 100 L, and 200 L)
  • Established products with set specifications are made to order
  • Product standardization enables shorter times from inquiry to quote and shipment, compared to custom
  • Readily accessible documentation for accelerated qualification

Benefits

  • Reduces time spent preparing buffers in-house
  • Mitigates space requirement for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Meets workflow needs across stages of development and manufacturing of biologicals
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
ColorClear, colorless solution
FormatSample
Manufacturing QualitycGMP
Osmolality265 to 295 mOsm/kg
Quantity5 L
Shelf Life24 Months
FormLiquid
Product TypeHanks' Balanced Salt Solution (HBSS)
SterilitySterile-filtered
Without AdditivesNo Calcium, No Magnesium
pH6.7 to 7.8
Unit SizeEach