Glucose, high-temperature short-time (HTST)-treated
Gibco™

Glucose, high-temperature short-time (HTST)-treated

Gibco™ high-temperature short-time (HTST)-treated products are briefly exposed to temperatures of ≥100°C using in-line pasteurization technology.
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerMengeKonzentrationFormat
A59774BD100 l in BioProcess-Container40% (w/v)trommel
A59773BA5l im BPC;25 % (G/V)Probe
A59773BB20 l im BPC25 % (G/V)Aufbewahrungsbehälter
A59773BC50l im BPC;25 % (G/V)trommel
A59773BD100 l in BioProcess-Container25 % (G/V)trommel
A59773BE200 l in BioProcess-Container25 % (G/V)trommel
A59774BA5l im BPC;40% (w/v)Probe
A59774BB20 l im BPC40% (w/v)Aufbewahrungsbehälter
A59774BC50l im BPC;40% (w/v)trommel
A59774BE200 l in BioProcess-Container40% (w/v)trommel
A59343BA5l im BPC;50 % (w/v)Probe
A59343BB20 l im BPC50 % (w/v)Aufbewahrungsbehälter
A59343BC50l im BPC;50 % (w/v)trommel
A59343BD100 l in BioProcess-Container50 % (w/v)trommel
A59343BE200 l in BioProcess-Container50 % (w/v)trommel
Katalognummer A59774BD
Preis (EUR)
-
Menge:
100 l in BioProcess-Container
Konzentration:
40% (w/v)
Format:
trommel
Gibco™ high-temperature short-time (HTST)-treated products are briefly exposed to temperatures of ≥100°C using in-line pasteurization technology. HTST is risk mitigation step that may reduce potential viral contaminations from entering upstream manufacturing from raw materials. Products such as glucose and other simple sugars are heated for a controlled period to not alter the treated product’s performance or viability. If HTST is a critical step in your process, outsourcing glucose can help you discover new efficiencies to reclaim time, space, and resources.

Product Features

  • Multiple formulations and concentrations available
  • Available in a range of pack sizes (5 L, 20 L, 50 L, 100 L, and 200 L)
  • Established products with set specifications are made to order
  • Product standardization enables shorter times from inquiry to quote and shipment, compared to custom
  • Readily accessible documentation for accelerated qualification

Benefits

  • Reduces time spent preparing buffers in-house
  • Mitigates space requirement for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Meets workflow needs across stages of development and manufacturing of biologicals
Für die Verwendung in der Forschung oder die Weiterverarbeitung. Nicht zur Diagnosestellung oder direkten Anwendung bei Menschen oder Tieren geeignet.
Specifications
FarbeClear, colorless solution
Konzentration40% (w/v)
Endotoxin-Gehalt≤1.00
Formattrommel
ZusammensetzungGlucose, HTST-treated, 40% (w/v)
Fertigungsqualität, HerstellungsqualitätcGMP
Menge100 l in BioProcess-Container
Haltbarkeit24 Monate
Getestet aufGlucose concentration
FormFlüssig
ProdukttypGlukose
SterilitätSteril gefiltert
Unit SizeEach