Fluorescein/Oregon Green Monoclonal Antibody (4-4-20) - FAQs
Fluorescein/Oregon Green Monoclonal Antibody (4-4-20) - FAQs
View additional product information for Fluorescein/Oregon Green Monoclonal Antibody (4-4-20) - FAQs (A6421)
1 product FAQs found
I want to label F-actin with phalloidin and image with transmission electron microscopy. What is the best way to go about it?
One way is to use a biotin-phalloidin conjugate, then label that with a colloidal gold conjugate of streptavidin. You will need to block endogenous biotin first. Another method is to label with fluorescein phalloidin, then with an anti-fluorescein antibody conjugated with colloidal gold.