Residual Protein TrypLE™ ELISA Kit, 10 kits, 960 tests - FAQs
Residual Protein TrypLE™ ELISA Kit, 10 kits, 960 tests - FAQs
View additional product information for Residual Protein TrypLE™ ELISA Kit - FAQs (A6533103, A6533101, A6533102)
1 product FAQs found
How can I assess residual TrypLE reagent remaining after my downstream processing steps?
We offer the Residual TrypLE ELISA Kit (Cat. No. A6533101) that can specifically detect the recombinant protease (rProtease) in TrypLE reagent.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.