POROS™ CaptureSelect™ C1-InhXL Affinity Resin
Thermo Scientific™

POROS™ CaptureSelect™ C1-InhXL Affinity Resin

Thermo Scientific™ POROS™ CaptureSelect™ C1-InhXL Affinity Resin is intended for the purification of human C1 esterase inhibitor from recombinant and plasma/serum sources.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A6543750 mL
A6543625 mL
A65439250 mL
A654401 L
A654415 L
A6544210 L
Catalog number A65437
Price (USD)
6,610.00
Each
Quantity:
50 mL
Price (USD)
6,610.00
Each

POROS CaptureSelect C1-InhXL Affinity Resin is intended for the purification of human C1 esterase inhibitor from recombinant and plasma/serum sources. C1 esterase inhibitor is a protease inhibitor belonging to the serpin superfamily that inhibits the spontaneous activation of the complement system. The CaptureSelect C1-InhXL affinity ligand is highly selective for human C1 inhibitor and coupled to the high-performing POROS resin, allowing for an efficient and scalable purification process, with high purity and yield obtained in a single step. The large pore structure of the POROS beads enables higher flowrates and efficient binding of larger biomolecules.

Features of POROS CaptureSelect C1-InhXL Affinity Resin include:

  • Efficient purification of C1 esterase inhibitor with high yield
  • Mild pH elution condition to retain activity
  • High binding capacity at high flow rates
  • Excellent scalability
  • Non-animal derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Resin: cross-linked poly(styrene-divinylbenzene) POROS beads
  • Average particle size: 50 μm
  • Ligand: single-domain [VHH] antibody fragment
  • Binding capacity: 20–25 g C1-Inhibitor/L resin
  • Elution conditions: 50 mM sodium acetate, 1.0 M MgCl2, pH 5.0
  • Regeneration conditions: acidic stripping condition with 0.5 M acetic acid, pH 2
  • Mechanical resistance: 100 bar (1450 psi, 10 MPa)
  • Formulation buffer: 18% (v/v) ethanol
For research and development use and further manufacturing in support of FDA-regulated end uses. Not for diagnostic use or direct administration to humans or animals.
Specifications
For Use With (Application)Protein purification
Quantity50 mL
Shipping ConditionWet Ice
TargetC1 Esterase Inhibitor
Binding Capacity20 to 25 g C1-Inhibitor/L Resin
FormResin Slurry
Particle Size50 μm
Product LinePOROS CaptureSelect
TypeAffinity Resin
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.