JUN™ Dye Spectral Calibration Plate for Multiplex qPCR, Fast 96-well
Applied Biosystems™

JUN™ Dye Spectral Calibration Plate for Multiplex qPCR, Fast 96-well

A specialized calibration plate containing JUN dye that is designed for use with the 7500 Fast Dx instrument and is utilized to perform spectral calibration and help ensure that the instrument maintains accurate spectral calibration information.
Have Questions?
Catalog number A65488
Price (USD)
178.00
Each
Price (USD)
178.00
Each

The JUN Dye Spectral Calibration Plate for Multiplex qPCR, Fast 96-well, is a specialized calibration plate containing JUN dye that is designed for use with the 7500 Fast Dx instrument and is utilized to perform spectral calibration and help ensure that the instrument maintains accurate spectral calibration information, which is crucial for reliable multiplex qPCR results.

This calibration plate is intended for in vitro diagnostic use by trained personnel in a professional environment to perform dye calibration of the 7500 Fast Dx real-time PCR instrument. For information about instrument setup and calibration, refer to the instrument user guide or contact Support.

For In Vitro Diagnostic Use. Not available in all countries or regions.
Specifications
Calibration TypeJUN™
Detection MethodFluorescence
DimensionsReal Time PCR (qPCR)
Display Type7500 Fast Dx
Dynamic RangeDye Spectral Calibration Plate
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)7500 Fast Dx
Format1 x 96-well plate
Quantity2 Plate
Shipping ConditionDry Ice
Product TypeDye Spectral Calibration Plate
Unit SizeEach
Contents & Storage
Store at -20°C ±5°C.