CLA IdentiFiler™ Plus PCR Amplification Kit
Applied Biosystems™

CLA IdentiFiler™ Plus PCR Amplification Kit

The CLA (Cell Line Authentication) IdentiFiler Plus PCR Amplification Kit simplifies the amplification step during authentication of cell lines.
Have Questions?
Catalog NumberQuantity
A65672200 x 25 μL Reactions
Catalog number A65672
Price (USD)
7,830.00
Each
Quantity:
200 x 25 μL Reactions
Price (USD)
7,830.00
Each

The CLA (Cell Line Authentication) IdentiFiler Plus PCR Amplification Kit simplifies the amplification step during authentication of cell lines. It includes the same primers and allelic ladder as the widely used AmpFLSTR Identifiler Plus kit and harnesses next-generation PCR amplification technology to help provide enhanced sensitivity, a cleaner baseline, improved performance with mixtures, and the ability to overcome high levels of PCR inhibition. The kit includes GeneScan 600 LIZ Size Standard v2.0, a five-dye-labeled, high-density size standard for the reproducible sizing of fragment analysis data.

Features of the CLA IdentiFiler Plus PCR Amplification Kit include:

  • 5-dye, 16-locus STR kit compatible with all cell line databasing loci standards
  • Improved version of legacy AmpFLSTR Identifiler Plus kit
  • Size standard included for ease of purchase
  • Compatible with SeqStudio, 3730/xL, and 3500/xL genetic analyzers

Cell line authentication using short tandem repeat (STR) analysis kits can be used both for human sample authentication (HSA) and mixed sample analysis (MSA). In HSA, there is a need to verify the DNA profile of the sample to check for sample cross-contamination or sample mix-up, or to perform a quality check. For MSA there is a need to deconvolute the presence of multiple contributors in a sample (chimerism) through identification of multiple DNA genotypes in the sample.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Equipment)SeqStudio Flex 3500, SeqStudio 3730xl Genetic Analyzers
FormatKit
Kit TypeMulticomponent
PolymeraseAmpliTaq Gold DNA Polymerase
Purification TargetCell DNA
Quantity200 x 25 μL Reactions
Reaction FormatTube
Sample TypeGenomic DNA
SpeciesHuman
Starting MaterialCell Line Culture
Target Organism ClassHuman cell lines
TechniqueSTR (Short Tandem Repeats) Analysis
Detection MethodGel Electrophoresis
For Use With (Application)Genotyping, Cell Analysis
FormLiquid
GC-Rich PCR PerformanceHigh
Label or DyeLIZ, FAM, NED, PET, VIC
PCR MethodEnd-point PCR
Reaction Speed60 min.
Unit SizeEach
Contents & Storage
• AmpFLSTR Identifiller Plus PCR Ampl Kit
• GS Liz600 Size Standard