Tris Base Solutions
Gibco™

Tris Base Solutions

Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidadConcentración
A65690BC50 L2 M
A65690BA5 L2 M
A65690BB20 L2 M
A65690BD100 L2 M
A65690BE200 L2 M
A65784BA5 L1 M
A65784BB20 L1 M
A65784BC50 L1 M
A65784BD100 L1 M
A65784BE200 L1 M
Número de catálogo A65690BC
Precio (MXN)
-
Cantidad:
50 L
Concentración:
2 M

Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.

Features

  • Concentrated for dilution on-site
  • Available in a range of pack sizes
  • Established shelf life: set specifications
  • Made to order
  • Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
  • Readily accessible documentation to help accelerate qualification

Benefits

  • Helps ease time requirements for in-house buffer preparation
  • Mitigates space constraints for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Addresses workflow needs for biologics development and manufacturing stages

Additional information

  • Multi-compendial testing: compendial-grade or equivalent raw materials
  • Primary packaging: TAegis 5-14 single-use bioprocess container (BPC)
  • Secondary packaging: rigid shipping container with appropriate dunnage
  • Manufacturing facilities: ISO 13485–certified, cGMP-compliant to 21 CFR Part 820 standard
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
CAS77-86-1
Calidad de fabricaciónCGMP
EnvaseBioprocess Container in Secondary Packaging
Concentración2 M
Para utilizar con (aplicación)Bioproduction
Para utilizar con (equipo)ILC/ILD System
GradoEP/USP
Peso molecular (g/mol)Raw Material: 121.14
Porcentaje de purezaNot Available
Forma físicaSolution
Cantidad50 L
Tipo de soluciónBuffer
pH10.0 to 12.0
Unit SizeEach