Hepes Solutions
Gibco™
Hepes Solutions
Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidadConcentración
A65782BD100 L2 M
A65782BA5 L2 M
A65782BB20 L2 M
A65782BC50 L2 M
A65782BE200 L2 M
A65783BA5 L1 M
A65783BB20 L1 M
A65783BC50 L1 M
A65783BD100 L1 M
A65783BE200 L1 M
Número de catálogo A65782BD
Precio (USD)
-
Cantidad:
100 L
Concentración:
2 M
Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.
Features
  • Concentrated for dilution on-site
  • Available in a range of pack sizes
  • Established shelf life: set specifications
  • Made to order
  • Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
  • Readily accessible documentation to help accelerate qualification
Benefits
  • Helps ease time requirements for in-house buffer preparation
  • Mitigates space constraints for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Addresses workflow needs for biologics development and manufacturing stages
Additional information
  • Multi-compendial testing: compendial-grade or equivalent raw materials
  • Primary packaging: TAegis 5-14 single-use bioprocess container (BPC)
  • Secondary packaging: rigid shipping container with appropriate dunnage
  • Manufacturing facilities: ISO 13485–certified, cGMP-compliant to 21 CFR Part 820 standard
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Concentración2 M
Calidad de fabricaciónCGMP
Cantidad100 L
FormularioClear, colorless solution
Tipo de productoHEPES Buffer
EsterilidadSterile-filtered
Sterilization MethodSterile-filtered
Unit SizeEach