POROS™ GoPure™ Oligo (dT)25 Screening Plate, 96-well, 20 μL
Thermo Scientific™

POROS™ GoPure™ Oligo (dT)25 Screening Plate, 96-well, 20 μL

Thermo Scientific™ POROS™ GoPure™ Oligo (dT)25 96-well screening plates contain POROS Oligo (dT)25 Affinity Resin and provide a versatile platform for sample preparation, handling, and analysis.
Have Questions?
Catalog NumberQuantity
A658111 Plate
Catalog number A65811
Price (MXN)
-
Quantity:
1 Plate

POROS GoPure Oligo (dT)25 96-well screening plates contain POROS Oligo (dT)25 Affinity Resin and provide a versatile platform for sample preparation, handling, and analysis. They accommodate multiple samples simultaneously to achieve high throughput and data generation.

These pre-filled screening plates have the following features:

  • High-throughput screening capability enabling multiple conditions to be evaluated in a single plate experiment
  • 20 μL per well, helping preserve sample while maximizing data generation
  • Ready-to-use 96-well format for efficient evaluation of Thermo Scientific resins
For Research Use Only.
Specifications
Capacity (Metric)800 μL
Concentration220 μL Resin Slurry/Well
For Use With (Application)Chromatography
Format96-well Filter Plate
Ligand TypePoly-deoxythymidine (dT25-mer) ligand with Proprietary Linker
Quantity1 Plate
Sample Volume20 μL
Shipping ConditionWet Ice
FormResin Slurry
Particle Size50 μm
Product LinePOROS
TypeAffinity Screening Plate
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C. Do not freeze.