CLA IdentiFiler™ Direct PCR Amplification Kit
Applied Biosystems™

CLA IdentiFiler™ Direct PCR Amplification Kit

The CLA (Cell Line Authentication) IdentiFiler Direct PCR Amplification Kit simplifies the amplification step during authentication of cell lines.
Have Questions?
Catalog NumberQuantity
A65908200 x 25 μL Reactions
Catalog number A65908
Price (USD)
9,090.00
Each
Quantity:
200 x 25 μL Reactions
Price (USD)
9,090.00
Each

The CLA (Cell Line Authentication) IdentiFiler Direct PCR Amplification Kit simplifies the amplification step during authentication of cell lines. The kit enables direct amplification of the same 16 loci included in our AmpFLSTR Identifiler Direct kit, eliminating the need to perform DNA extraction or purification steps for DNA database and other single-source reference samples. Blood or buccal samples on FTA cards can be punched into PCR plates or tubes, and taken directly to PCR amplification, without reduction in data quality. Samples collected using a Buccal DNA Collector or other non-FTA substrate can be processed without heat incubation or additional workflow steps using Prep-n-Go Buffer (available separately). The CLA Identifiler kit includes GeneScan 600 LIZ Size Standard v2.0, a five-dye-labeled, high-density size standard for the reproducible sizing of fragment analysis data.

Features of the CLA IdentiFiler Direct PCR Amplification Kit include:

  • 5-dye, 16-locus STR analysis kit compatible with all cell line databasing loci standardsImproved version of legacy AmpFLSTR Identifiler Direct kit
  • Size standard included for ease of purchase
  • Compatible with SeqStudio, 3730/xL, and 3500/xL genetic analyzers

Cell line authentication using short tandem repeat (STR) analysis kits can be used both for human sample authentication (HSA) and mixed sample analysis (MSA). In HSA, there is a need to verify the DNA profile of the sample to check for sample cross-contamination or sample mix-up, or to perform a quality check. For MSA there is a need to deconvolute the presence of multiple contributors in a sample (chimerism) through identification of multiple DNA genotypes in the sample.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Equipment)SeqStudio Flex™ 3500, SeqStudio™ 3730xl Genetic Analyzers
FormatKit
Isolation TechnologyDirect PCR
Kit TypeMulticomponent
No. of Reactions200 x 25 μL Reactions
PolymeraseAmpliTaq Gold™ DNA Polymerase
Purification TargetGenomic DNA from human cells
Quantity200 x 25 μL Reactions
Reaction FormatMaster Mix
Sample TypeCell Line Culture
SpeciesHuman
Target Organism ClassCell lines
TechniqueSTR (Short Tandem Repeats) Analysis
Detection MethodGel Electrophoresis
For Use With (Application)Genotyping, Cell Analysis
FormLiquid
GC-Rich PCR PerformanceHigh
Label or DyeLIZ, FAM, NED, PET, VIC
PCR MethodDirect PCR
Reaction Speed60 min.
Unit SizeEach
Contents & Storage
• IdentiFiler Direct PCR Amplification Kit
• Liz600 Size Standard