ATTO-TAG™ FQ Plus Amine-Derivatization Kit - FAQs
ATTO-TAG™ FQ Plus Amine-Derivatization Kit - FAQs
View additional product information for ATTO-TAG™ FQ Plus Amine-Derivatization Kit - FAQs (A66521)
4 product FAQs found
Can I use the ATTO-TAG FQ Plus Amine Derivatization Kit to label small peptides?
Yes, the ATTO-TAG FQ Plus Amine Derivatization Kit is suitable for labeling proteins, peptides, and amino sugars.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
The ATTO-TAG FQ Plus Amine Derivatization Kit was shipped on wet ice but the storage recommendation is at -20 degrees C? Is it still good to use?
Yes, the product is still good to use. We ship many of our products at a higher temperature than the storage recommendation. The product will be stable for at least one week at the shipping temperature when in the original, unopened packaging. For long-term stability, the product should be placed at the recommended storage temperature upon receipt.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
What is the difference between the ATTO-TAG FQ Plus Amine Derivatization Kit (Cat. No. A66521) and the ATTO-TAG FQ Amine-Derivatization Kit (Cat. No. A2334)?
The ATTO-TAG FQ Amine Derivatization Kit (Cat. No. A2334) contains a potassium cyanide (KCN) component (component B). This component has been removed in the ATTO-TAG FQ Plus Amine Derivatization Kit (Cat. No. A66521) and replaced with mandelonitrile which serves as a less toxic alternative to KCN without compromising assay performance.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
Where can I find the expiration date for the ATTO-TAG FQ Plus Amine Derivatization Kit?
The ATTO-TAG FQ Plus Amine Derivatization Kit does not have an expiration date. The kit is guaranteed for 12 months from date of shipment, when stored as indicated, in the original unopened packaging. Stock solutions should be stable for 6 months after reconstitution of the ATTO-TAG FQ (component A) provided good quality methanol is used for reconstitution (methanol not included in the kit).
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.