Bacto™ CD Supreme Fermentation Production Medium Plus
Gibco™
Bacto™ CD Supreme Fermentation Production Medium Plus
Chemically defined medium optimized for E. coli plasmid DNA and recombinant protein production.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
A937850210 kg
A9378501500 g
Número de catálogo A9378502
Precio (USD)
-
Cantidad:
10 kg
Gibco Bacto CD Supreme Fermentation Production Medium (FPM) Plus is the next-generation, chemically defined formulation designed to support a wide range of Escherichia coli (E. coli) strains used in plasmid DNA and recombinant protein production. This hydrolysate-free medium enhances microbial culture performance in both batch and fed-batch fermentation processes, offering superior consistency and flexibility across manufacturing scales.
Features of Bacto CD Supreme FPM Plus include:
  • Consistency with a 100% chemically defined, hydrolysate-free formulation that minimizes lot-to-lot variability
  • Expanded strain compatibility to include E. coli strains such as DH10B, BL21(DE3), NEB™ Stable, Stbl2, Stbl3, JM109, and other auxotrophic variants
  • Simplified workflows using a one-part, ready-to-use formulation that eliminates manual media preparation
  • Easy adaptation to your process with format options and compatibility with filtration or autoclave sterilization methods
  • Reduced regulatory risk with a formulation free from raw materials of animal origin or materials produced using animal-derived components
Bacto CD Supreme FPM Plus is engineered to deliver reliable and reproducible growth performance across diverse E. coli host strains, including Thi-1 mutant and auxotrophic variants. Its chemically defined composition helps ensure optimal nutrient availability without the variability introduced by hydrolysates. The medium supports high-density cultures in both batch and fed-batch modes, enabling efficient plasmid DNA or recombinant protein yield.
For fed-batch processes Bacto CD Supreme FPM Plus can be combined with Bacto CD Supreme Feed (Cat. Nos. A9378201, A9378301, A9378302) to support increased biomass accumulation and yields
This one-part, easy-to-use formulation integrates smoothly into existing manufacturing workflows and is compatible with standard sterilization methods. It is produced without any materials of animal origin, minimizing regulatory complexity and eliminating TSE/BSE risk concerns.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Para utilizar con (aplicación)Microbial Fermentation
FormulaciónAnimal Origin-free, Chemically-defined
Línea de productosBacto CD Supreme
Cantidad10 kg
Duración de almacenamiento12 Months
FormularioDehydrated Powder
Origen de fabricaciónUnited States
Tipo de productoCulture Medium
EsterilidadNon-sterile
Clase de organismo dianaE. coli
Tipo de pruebaMicrobial Cultivation
Unit SizeEach
Contenido y almacenamiento
Store in the dark at +2°C to +8°C