Tubes, 0.5 mL, domed cap, assorted colors - FAQs
Tubes, 0.5 mL, domed cap, assorted colors - FAQs
View additional product information for Tubes, 0.5 mL, domed cap, assorted colors - FAQs (AB0535)
1 product FAQs found
Which plastic consumables should I use on my real-time PCR (qPCR)/thermal cycler (PCR) instrument?
Please select your real-time PCR (qPCR) or thermal cycler (PCR) instrument model from the drop-down menu in the qPCR and PCR Plastics Selection Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html) to see the list of all validated plastic consumables (plates, tube strips, single tubes, caps, seals, adhesive films, and accessories) we offer that are compatible with your instrument.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.