ALPS Heat Sealer Accessories
ALPS Heat Sealer Accessories
ALPS Heat Sealer Accessories
ALPS Heat Sealer Accessories
Thermo Scientific™
ALPS Heat Sealer Accessories
Use these ALPS heat sealer accessories with the ALPS 25 Manual Heat Sealer and ALPS 50 V Microplate Heat Sealer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
AB-05631000
also known as AB-0563/1000
96-Well PCR Plate Carrier
AB-0724384-well PCR Plate Carrier
AB-0827Standard Microplate Carrier
AB-1118V Bottom 96-Well Storage Plate Carrier
AB-1177ALPS 25 Sealer Stand and Shuttle Assembly Upgrade
Catalog number AB-05631000
also known as AB-0563/1000
Price (USD)
437.40
Online Exclusive
510.00
Save 72.60 (14%)
Each
Description:
96-Well PCR Plate Carrier
Price (USD)
437.40
Online Exclusive
510.00
Save 72.60 (14%)
Each
Use Thermo Scientific™ ALPS heat sealer accessories with the ALPS 25 Manual Heat Sealer and ALPS 50 V Microplate Heat Sealer. The foil stripper, AB-0592, is also compatible with the ALPS 3000 Microplate Heat Sealer.
 Warranty: 1 year
Specifications
Description96-Well PCR Plate Carrier
For Use With (Equipment)ALPS™ Heat Sealer
No. per Pack1 Ea.
Product TypeCarrier
Unit SizeEach