Easy Pierce Heat Sealing Foil
Actual product may vary
Thermo Scientific™

Easy Pierce Heat Sealing Foil

Save time and costs while improving sealing integrity with these microplate heat sealing instruments. Offers the capability of retrieving samples and then re-sealing the plate.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialProduct TypeDimensions (L x W)
AB-0757AluminumEasy Pierce Foil85 x 135 mm
AB-1797PolyesterClear Seal Thin80 x 125 mm
AB-0812PolyesterDiamond Clear Seal85 x 135 mm
AB-0559AluminumThermo-Seal85 x 135 mm
Catalog number AB-0757
Price (USD)
141.27
Online exclusive
156.83
Save 15.56
Case of 100
Material:
Aluminum
Product Type:
Easy Pierce Foil
Dimensions (L x W):
85 x 135 mm
Price (USD)
141.27
Online exclusive
156.83
Save 15.56
Case of 100

Save time and costs while improving sealing integrity with Thermo Scientific™ Heat Seals for ALPS™ Microplate Heat Sealing Instruments. Ideal for storage applications, heat sealing offers the capability of retrieving samples and then re-sealing the plate. When heat sealing, the sealing material is first placed in position over the plate, which must have a raised rim around each well. Heat is applied evenly for several seconds, resulting in binding of the sealing material to the raised rim to achieve a complete seal.

Features:

  • Quick operation
  • Safe for temperature-sensitive samples
  • Designed for reliable performance with Thermo Scientific ALPS instruments, including the, ALPS 3000, ALPS 25 and ALPS 50 V
Specifications
AutoclavableNot Autoclavable
Dimensions (L x W)85 x 135 mm
For Use With (Application)PCR, Ideal for compound management and high throughput screening applications
For Use With (Equipment)ALPS 25 and ALPS 50V, AB-1443A
Incubation ConditionsGeneral
PeelableYes
Product TypeEasy Pierce Foil
Quantity100 Pk.
Solvent Or Alcohol ResistantLife Science Reagents Resistant, DMSO Resistant
Temperature Range (Metric)-80°C to +80°C, -80°C to +120°C (with Adequate Pressure)
Well ShapeAll well shapes
AdhesivePolypropylene
Certified RNase/DNase-freeNo
ColorFoil
MaterialAluminum
No. of Wells96
PierceableYes
Plate CompatibilityPolypropylene, Polyethylene
Product LineAbgene
SterilityNon-sterile
Unit SizeCase of 100
Contents & Storage
General, Long Term