Easy Pierce Heat Sealing Foil
Actual product may vary
Thermo Scientific™

Easy Pierce Heat Sealing Foil

Save time and costs while improving sealing integrity with these microplate heat sealing instruments. Offers the capability of retrieving samples and then re-sealing the plate.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialProduct TypeDimensions (L x W)
AB-0812PolyesterDiamond Clear Seal80 x 125 mm
AB-1797PolyesterClear Seal Thin80 x 125 mm
AB-0559AluminumThermo-Seal85 x 135 mm
AB-0757AluminumEasy Pierce Foil85 x 135 mm
Catalog number AB-0812
Price (MXN)
-
Material:
Polyester
Product Type:
Diamond Clear Seal
Dimensions (L x W):
80 x 125 mm

Save time and costs while improving sealing integrity with Thermo Scientific™ Heat Seals for ALPS™ Microplate Heat Sealing Instruments. Ideal for storage applications, heat sealing offers the capability of retrieving samples and then re-sealing the plate. When heat sealing, the sealing material is first placed in position over the plate, which must have a raised rim around each well. Heat is applied evenly for several seconds, resulting in binding of the sealing material to the raised rim to achieve a complete seal.

Features:

  • Quick operation
  • Safe for temperature-sensitive samples
  • Designed for reliable performance with Thermo Scientific ALPS instruments, including the, ALPS 3000, ALPS 25 and ALPS 50 V
Specifications
Dimensions (L x W)80 x 125 mm
FluorescenceLow Autofluorescence
For Use With (Application)Ideal for qPCR and other fluorescence, colorimetric, and optical applications, suitable for PCR, ELISA
For Use With (Equipment)ALPS 25 and ALPS 50V, AB-1443A, Recommended for sealing Fast Diamond 384 PCR plates
Incubation ConditionsGeneral
PeelableYes
Product TypeDiamond Clear Seal
Quantity100 Pk.
Solvent Or Alcohol ResistantSome Solvent Resistance
Temperature Range (Metric)-80°C to +120°C
AdhesivePolypropylene
ColorClear
MaterialPolyester
PierceableNo
Plate CompatibilityPolypropylene, Polyethylene, Cyclic Olefin Copolymer, Polystyrene
Product LineAbgene
Unit SizeCase of 100
Contents & Storage
General, Long Term