Easy Pierce Heat Sealing Foil
Actual product may vary
Thermo Scientific™

Easy Pierce Heat Sealing Foil

Save time and costs while improving sealing integrity with these microplate heat sealing instruments. Offers the capability of retrieving samples and then re-sealing the plate.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialProduct TypeDimensions (L x W)
AB-1797PolyesterClear Seal Thin80 x 125 mm
AB-0812PolyesterDiamond Clear Seal85 x 135 mm
AB-0559AluminumThermo-Seal85 x 135 mm
AB-0757AluminumEasy Pierce Foil85 x 135 mm
Catalog number AB-1797
Price (USD)
1,282.28
Online exclusive
1,496.50
Save 214.22
Case of 1000
Material:
Polyester
Product Type:
Clear Seal Thin
Dimensions (L x W):
80 x 125 mm
Price (USD)
1,282.28
Online exclusive
1,496.50
Save 214.22
Case of 1000

Save time and costs while improving sealing integrity with Thermo Scientific™ Heat Seals for ALPS™ Microplate Heat Sealing Instruments. Ideal for storage applications, heat sealing offers the capability of retrieving samples and then re-sealing the plate. When heat sealing, the sealing material is first placed in position over the plate, which must have a raised rim around each well. Heat is applied evenly for several seconds, resulting in binding of the sealing material to the raised rim to achieve a complete seal.

Features:

  • Quick operation
  • Safe for temperature-sensitive samples
  • Designed for reliable performance with Thermo Scientific ALPS instruments, including the, ALPS 3000, ALPS 25 and ALPS 50 V
Specifications
AutoclavableNot Autoclavable
Dimensions (L x W)80 x 125 mm
FluorescenceLow Autofluorescence
For Use With (Application)Ideal for fluorescence, colorimetric, and optical applications, suitable for PCR/ qPCR, ELISA
For Use With (Equipment)ALPS 3000, ALPS 25 and ALPS 50 V, AB-1443A
Incubation ConditionsGeneral
PeelableYes
Product TypeClear Seal Thin
Quantity1000 Pk.
Solvent Or Alcohol ResistantLife Science Reagent Resistant
Temperature Range (Metric)-80°C to +120°C
Well ShapeAll well shapes
AdhesivePolypropylene
Certified RNase/DNase-freeNo
ColorClear
MaterialPolyester
No. of Wells96
PierceableYes
Plate CompatibilityPolypropylene, Polyethylene, Polystyrene
Product LineAbgene
SterilityNon-sterile
Unit SizeCase of 1000
Contents & Storage
General, Long Term