Product Image
Thermo Scientific™
Oxoid™ Anaerobic 2.5 L Rectangular Jar Complete
Anaerobic 2.5 Litre Rectangular Jar Complete Stackable
Have Questions?
Catalog NumberQuantity
AB0025AEach
Catalog number AB0025A
Price (USD)
-
Quantity:
Each
Anaerobic 2.5 Litre Rectangular Jar Complete Stackable holds up to 12x 90mm plates or 21 x 55mm plates
Specifications
QuantityEach
TypeRectangular Jar
Unit SizeEach