Tubes, strips of 8, 10 Packs of 12 Strips - FAQs
Tubes, strips of 8, 10 Packs of 12 Strips - FAQs
View additional product information for Tubes, strips of 8 - FAQs (AB0452, AB0264)
1 product FAQs found
Are Thermo Scientific 0.2 mL Strip Tubes autoclavable?
Yes, our Thermo Scientific PCR plastics are autoclavable. Our recommendation is to autoclave for 15 min at 121 degrees C.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.